ContactUsLinkComponent
Shodex HILICpak VG-50 HILIC Columns for Polar Compounds and Sugar Analysis
Shodex HILICpak VG-50 HILIC Columns for Polar Compounds and Sugar Analysis
Supplier:  Resonac
Certificates
undefined
Shodex HILICpak VG-50 HILIC Columns for Polar Compounds and Sugar Analysis
Supplier:  Resonac
Specifications
  • Catalog No:
  • SHOWF7630300
  • SHOWF6711200
  • 554-4573
  • 554-4572
  • 554-4574
  • Description:
  • HILICpak VG-50 2D
  • HILICpak VG-50G 2A
  • HILICpak VG-50 4D
  • HILICpak VG-50 4E
  • HILICpak VG-50G 4A
  • Max. pressure:
  • 15 MPa (150 bar)
  • 10 MPa (100 bar)
  • 15 MPa (150 bar)
  • 150 bar
  • pH range:
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • Temperature limit:
  • 4 - 60 °C
  • 4 - 60 °C
  • 4 - 60 °C
  • 4 - 60 °C
  • 4 - 60 °C