Cooling water tubing
Lieferant: LAUDA
Zertifikate
Über diesen Artikel
EPDM tubing, reinforced.
Special polymer tubings for high pressure. Not suitable for Ultra 350 and mineral oils.
Frequently Bought Together
EPDM tubing, reinforced.
Special polymer tubings for high pressure. Not suitable for Ultra 350 and mineral oils.