Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Storage Boxes

Storage Boxes
 
Storage Boxes
Storage Boxes

 

 

Box in bright colours made of PP for cell culture tubes of 15 or 50 ml (Falcon). Two grids guarantee a safe stand of the tubes in the upper as well as the lower area.

  • Clear lid is providing the correct view
  • Temperature resistant from −80 °C up to +121 °C
  • Autoclavable
Product Family Options

For 15 ml tubes

 

  • No. of Holes
    Colour
    For tubes
     
     
  • 25 (5×5)
    Transparent
    15 ml (Ø 17 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 16,90 €
    16,90 €
    1 * 1 ST
       16,90 €

  • 25 (5×5)
    Yellow
    15 ml (Ø 17 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 19,00 €
    19,00 €
    1 * 1 ST
       19,00 €

  • 25 (5×5)
    Pink
    15 ml (Ø 17 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 17,80 €
    17,80 €
    1 * 1 ST
       17,80 €

  • 25 (5×5)
    Green
    15 ml (Ø 17 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 16,90 €
    16,90 €
    1 * 1 ST
       16,90 €

  • 25 (5×5)
    Blue
    15 ml (Ø 17 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 17,70 €
    17,70 €
    1 * 1 ST
       17,70 €

  • 25 (5×5)
    Orange
    15 ml (Ø 17 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 16,90 €
    16,90 €
    1 * 1 ST
       16,90 €

For 50 ml tubes

 

  • No. of Holes
    Colour
    For tubes
     
     
  • 9 (3×3)
    Transparent
    50 ml (Ø 30 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 17,80 €
    17,80 €
    1 * 1 ST
       17,80 €

  • 9 (3×3)
    Yellow
    50 ml (Ø 30 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 24,50 €
    24,50 €
    1 * 1 ST
       24,50 €

  • 9 (3×3)
    Pink
    50 ml (Ø 30 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 24,00 €
    24,00 €
    1 * 1 ST
       24,00 €

  • 9 (3×3)
    Green
    50 ml (Ø 30 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 17,80 €
    17,80 €
    1 * 1 ST
       17,80 €

  • 9 (3×3)
    Blue
    50 ml (Ø 30 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 18,50 €
    18,50 €
    1 * 1 ST
       18,50 €

  • 9 (3×3)
    Orange
    50 ml (Ø 30 mm)
       
       
    1 * 1 ST
    1 * 1 ST - 24,00 €
    24,00 €
    1 * 1 ST
       24,00 €