ContactUsLinkComponent
Syringe for vapour pressure osmometer
Syringe for vapour pressure osmometer
Catalog # KNAUA0433
Supplier:  KNAUER
Syringe for vapour pressure osmometer
Catalog # KNAUA0433
Supplier:  KNAUER
Specifications
  • Volume:
    1000 µl
  • Description:
    Syringe with stainless steel piston
  • Pk:
    1
Specifications
About this item
This syringe has a glass barrel, stainless steel piston with PTFE seal, including needle with spring stopper.