Quality control test suspensions
Supplier: CROSSTEX
Certificates
About this item
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.
- 10 ml pro vial
- 100 dose
Frequently Bought Together
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.