Standard mantory signs
: Brady
Mantory signs have always a blue circular backround which contains a white pictogram.These signs display a message or action that must be carried out in a designated area.
- Laminated polyester
- Resistance to most chemical products and solvents
- Resistance against extreme temperatures
- Good weatherability and UV resistance
- Excellent adhesion to low surface energy surfaces
- Thickness: 0,1 mm
- Different sizes
Frequently Bought Together