Order Entry
Austria
ContactUsLinkComponent
Worktop trolleys
Worktop trolleys
Catalog # SOVEC70141001
Supplier:  TRESTON
Worktop trolleys
Catalog # SOVEC70141001
Supplier:  TRESTON

Specifications

  • Description:
    Worktop trolley, height adjustable from 655 - 1055 mm
  • W×D×H:
    750×600×655/1055 mm
  • Pk:
    1

Specifications

About this item

The Worktop trolleys are light duty trolleys which provides mobile workspace and are ideal for use in industries.

  • Equipped with four swivel casters of Ø 100 mm and two of which have brakes.
  • Load capacity: 150 kg