Order Entry
Austria
ContactUsLinkComponent
 
Anti-Histone H3 Rabbit Polyclonal Antibody
  BSENR-1549-100
 :  Biosensis
undefined
Anti-Histone H3 Rabbit Polyclonal Antibody
  BSENR-1549-100
 :  Biosensis

 

  • Antibody type:
    Primary
  • Antigen name:
    Histone H3.1
  • Antigen symbol:
    Histone H3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminus of Human Histone H3 (121-136 aa).
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    A peptide from the C-terminus of human Histone H3 (121-136 aa).
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Histone H3 is a core component of nucleosome.

Antibody binds to various Histone H3 proteins.
Application Information:
WB. Suggested dilution of 1:1,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Human Histone H3 (121-136 aa).
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine