Order Entry
Austria
ContactUsLinkComponent
Anti-Atrial Natriuretic Peptide Rabbit Polyclonal Antibody
Anti-Atrial Natriuretic Peptide Rabbit Polyclonal Antibody
Catalog # BSENR-1500-50
Supplier:  Biosensis
undefined
Anti-Atrial Natriuretic Peptide Rabbit Polyclonal Antibody
Catalog # BSENR-1500-50
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Natriuretic peptide A
  • Antigen symbol:
    NPPA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Epitope:
    rat Atrial natriuretic peptide (123-150 aa)
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Atrial natriuretic peptide|ANP
  • Immunogen:
    Synthetic rat Atrial natriuretic peptide (123-150 aa) conjugated to BSA.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

The hormone Atrial natriuretic peptide (ANP) is produced by cardiomyocites and is secreted into the bloodstream. ANP acts on convoluted tubules in kidney reducing sodium absorption which results in reducing the blood pressure.

The antibody recognizes ANP by native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity: Human; mouse; rat. Highly conserved and expected to interact with feline, canine, porcine, bovine, and equine ANP.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Atrial Natriuretic Peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: rat Atrial natriuretic peptide (123-150 aa)
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat