Order Entry
Austria
ContactUsLinkComponent
 
Anti-Nicastrin, N-terminal domain Rabbit Polyclonal Antibody
  BSENR-1686-500
 :  Biosensis
undefined
Anti-Nicastrin, N-terminal domain Rabbit Polyclonal Antibody
  BSENR-1686-500
 :  Biosensis

 

  • Antibody type:
    Primary
  • Antigen name:
    Nicastrin, N-terminal domain
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    Human Nicastrin [32-44]
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Immunogen:
    A synthetic peptide (RGNSVERKIYIPL-C) corresponding to human Nicastrin [32-44] in the N-terminal region conjugated via additional C-terminal Cys to Diphtheria toxoid.
  • Size:
    500 µg
  • Pk:
    500 µG

 

 

Nicastrin, a type 1 membrane glycoprotein, is an essential component of the gamma secretase complex which is critical for the cleavage of the amyloid precursor protein and other membrane proteins. Nicastrin is widely expressed in different tissue types. This antibody detects all processed forms of Nicastrin.

Confirmed by WB using peptide absorption.
Application Information:
WB and IP. Suggested concentration of 3-10 ug/mL is recommended for WB. Human or mouse brain samples commonly prepared with reducing (50mM DTT), urea (2.3 M), SDS (1.5%) in 62.5 mM Tris-HCL pH 6.8 sample buffer (without boiling) heating to 50 C for 15 min. Unprocessed full length human Nicastrin is 709 amino acids, however this protein contains an N-terminal signal peptide which is considered to undergo cleavage during processing and transit to the cell plasma membrane, in addition the protein undergoes glycosylation to produce a glycoprotein of about 145 kDa apparent MW by SDS PAGE. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Nicastrin, N-terminal domain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Nicastrin [32-44]
Host: Rabbit
Isotype: IgG
Reactivity: