Order Entry
Austria
ContactUsLinkComponent
Anti-beta synuclein Rabbit Polyclonal Antibody
Anti-beta synuclein Rabbit Polyclonal Antibody
Catalog # BSENR-1683-500
Supplier:  Biosensis
undefined
Anti-beta synuclein Rabbit Polyclonal Antibody
Catalog # BSENR-1683-500
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta synuclein
  • Antigen symbol:
    beta synuclein
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    C-terminal domain
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Immunogen:
    A synthetic peptide (AQEAAEEPLIEPLME-C) corresponding to human β-synuclein [99-113] in the C-terminal domain conjugated via additional C-terminal Cys to Diphtheria toxoid.
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Beta-synuclein is a soluble cytoplasmic protein associated with synaptic vesicles and a member of the synuclein family. Mutations in alpha-synuclein cause early onset Parkinson's disease. Expression of beta synuclein may modulate alpha-synuclein aggregation found in Parkinson's disease.

Confirmed by WB using soluble mouse and human brain extracts, reactivity for major product diminished by peptide absorption. Does not detect alpha-synuclein as tested with recombinant protein and does not react with Lewy bodies in human Dementia with Lewy Bodies or Parkinson's disease brain tissue sections.
Application Information:
WB and IHC. A dilution of 1:500 to 1:1,000 is recommended for Western blot. Beta-synuclein is a soluble protein of 134 amino acids and detected with 17 kDa mobility by western blotting. By IHC the antibody detects synaptic sites in human brain formaldehyde-treated frozen tissue. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: beta synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal domain
Host: Rabbit
Isotype: IgG
Reactivity: