Order Entry
Austria
ContactUsLinkComponent
Anti-Galectin-3 Mouse Monoclonal Antibody [clone: 5C21]
Anti-Galectin-3 Mouse Monoclonal Antibody [clone: 5C21]
Catalog # BSENM-1574-100
Supplier:  Biosensis
undefined
Anti-Galectin-3 Mouse Monoclonal Antibody [clone: 5C21]
Catalog # BSENM-1574-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Galectin 3
  • Antigen symbol:
    GAL3
  • Clonality:
    Monoclonal
  • Clone:
    5C21
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Carbohydrate-binding protein 35|Galactose-specific lectin 3|Macrophage galactose-specific lectin|CBP 35
  • Immunogen:
    Full length recombinant Galectin-3 expressed in and purified from E. coli.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Galectin 3 is a lectin with carbohydrate recognition domains (CRD) which bind  -galactoside. It is a multifunctional protein expressed both on the cell surface, cytoplasm and nucleus and appears to have roles in specific carbohydrate binding and in the regulation of mRNA splicing.

Antibody recognizes Galectin 3 by western blots (1:2000-1"10000) and in immunocytochemistry or Immunohistory chemistry on frozen or 4% PFA fixed tissues. Antibody has not been tested with paraffin tissues. Antibody reacts with human, mouse and rat and likely other species because of homology but this has not be confirmed.
Application Information:
WB, IHC/IF. Suggested dilution of at least 1:1,000 for IHC/IF. Suggested dilution of 1:10,000 or lower for WB.

Type: Primary
Antigen: Galectin-3
Clonality: Monoclonal
Clone: 5C21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat