Order Entry
Austria
ContactUsLinkComponent
Anti-Neurofilament Medium Mouse Monoclonal Antibody [clone: 3H11]
Anti-Neurofilament Medium Mouse Monoclonal Antibody [clone: 3H11]
Catalog # BSENM-1394-100
Supplier:  Biosensis
undefined
Anti-Neurofilament Medium Mouse Monoclonal Antibody [clone: 3H11]
Catalog # BSENM-1394-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurofilament Medium
  • Clonality:
    Monoclonal
  • Clone:
    3H11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Chicken,
    Human,
    Bovine,
    Rat,
    Mouse,
    Cat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Neurofilament medium polypeptide|Nfm,
  • Immunogen:
    Raised against a recombinant fusion protein containing the extreme C-terminus of rat NF-M expressed in and purified from E. coli. The epitope is localized to within the last 56 amino acids at the extreme C-terminus of rat NF-M, the so-called KE segment which is highly conserved between NF-M molecules from different species.
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Neurofilaments are composed of three intermediate filament proteins: light (~68 kDa), medium (~160 kDa) and heavy (~200 kDa), which are involved in the maintenance of the neuronal caliber. Neurofilament medium runs on SDS-PAGE gels in the range 145-170 kDa, with some variation in different species.

Specifically recognizes the medium neurofilament subunit NF-L in WB.
Application Information:
Western Blotting (WB), Immunocytochemistry (IC) and Immunohistochemistry (IH). A dilution of 1:1,000 - 1:5,000 is recommended for WB. A dilution of 1:100 - 1:500 is recommended for IC and IH. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Neurofilament Medium
Clonality: Monoclonal
Clone: 3H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse, Cat, Bovine, Pig, Chicken