Order Entry
Austria
ContactUsLinkComponent
Anti-C-reactive Protein terminal peptide Chicken Polyclonal Antibody
Anti-C-reactive Protein terminal peptide Chicken Polyclonal Antibody
Catalog # BSENC-1511-100
Supplier:  Biosensis
undefined
Anti-C-reactive Protein terminal peptide Chicken Polyclonal Antibody
Catalog # BSENC-1511-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-Reactive Protein
  • Antigen symbol:
    CRP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal region of Mouse CRP, amino acids 210-225.
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    A peptide from the C-terminus of mouse C-reactive protein (210-225 aa).
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

C-reactive protein has several roles associated with host defence such as; promoting agglutination, bacterial capsular swelling, phagocytosis and complement fixation through its calcium-dependent binding to phosphorylcholine. It can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells. COFACTOR: Binds 2 calcium ions per subunit. C-reactive protein exists as a homopentamer. There are 2 alternatively spliced isoforms. C-reactive protein is found in plasma and its concentration increases greatly during acute phase response to tissue injury, infection or other inflammatory stimuli. It is induced by IL-1 and IL-6.

The antibody detects mouse C-reactive protein in western blots by virtue of its epitope being the C-terminal peptide of the protein, amino acids 210-225. Because of homology, reactivity with C-terminal protein from other species is expected. The antibody also reacts in ELISA with mouse C-reactive protein
Application Information:
ELISA and WB. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: C-reactive Protein terminal peptide
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal region of Mouse CRP, amino acids 210-225.
Host: Chicken
Isotype: IgY
Reactivity: Mouse