Order Entry
Austria
ContactUsLinkComponent
Anti-Uncoupling Protein 3 Chicken Polyclonal Antibody
Anti-Uncoupling Protein 3 Chicken Polyclonal Antibody
Catalog # BSENC-1542-100
Supplier:  Biosensis
undefined
Anti-Uncoupling Protein 3 Chicken Polyclonal Antibody
Catalog # BSENC-1542-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Uncoupling protein 3
  • Antigen symbol:
    UCP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Pig,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminus of Human Uncoupling Protein 3 (298-312 aa).
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    A peptide from the C-terminus of human Uncoupling Protein 3 (298-312 aa).
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Uncoupling Protein 3 (UCP3) belongs to the mitochondrial carrier family. Located in the mitochondrion inner membrane, UCP3 creates proton leaks across the membrane thus uncoupling oxidative phosphorylation (Ref: SWISSPROT).

This antibody detects UCP3 at approx 35 kDa.
Application Information:
WB. Suggested dilution at 1:1,000 to 1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Uncoupling Protein 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Human Uncoupling Protein 3 (298-312 aa).
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat, Pig