Order Entry
Austria
ContactUsLinkComponent
Anti-C-Reactive Protein Chicken Polyclonal Antibody
Anti-C-Reactive Protein Chicken Polyclonal Antibody
Catalog # BSENC-1540-100
Supplier:  Biosensis
undefined
Anti-C-Reactive Protein Chicken Polyclonal Antibody
Catalog # BSENC-1540-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-Reactive Protein
  • Antigen symbol:
    CRP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Epitope:
    rat C-reactive protein (108-121 aa
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    A peptide from rat C-reactive protein (108-121 aa).
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

C-reactive protein has several roles associated with host defence such as; promoting agglutination, bacterial capsular swelling, phagocytosis and complement fixation through its calcium-dependent binding to phosphorylcholine. It can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells. COFACTOR: Binds 2 calcium ions per subunit. C-reactive protein exists as a homopentamer. There are 2 alternatively spliced isoforms. C-reactive protein is found in plasma and its concentration increases greatly during acute phase response to tissue injury, infection or other inflammatory stimuli. It is induced by IL-1 and IL-6.

Antibody recognizes C-reactive protein by western blot and ELISAs, other applications have not been tested.
Application Information:
WB and ELISA. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: C-Reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: rat C-reactive protein (108-121 aa
Host: Chicken
Isotype: IgY
Reactivity: Rat