Order Entry
Austria
ContactUsLinkComponent
Anti-V5 Tag Mouse Monoclonal Antibody [clone: BE10]
Anti-V5 Tag Mouse Monoclonal Antibody [clone: BE10]
Catalog # BSENM-1317-100
Supplier:  Biosensis
undefined
Anti-V5 Tag Mouse Monoclonal Antibody [clone: BE10]
Catalog # BSENM-1317-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    V5 tag
  • Antigen symbol:
    V5
  • Clonality:
    Monoclonal
  • Clone:
    BE10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Epitope:
    GKPIPNPLLGLDST
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    Synthetic peptide GKPIPNPLLGLDST
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The V5 epitope corresponds to a region from Simian Virus Type 5 (SV5).

Antibody detects V5-tagged fusion protein.
Application Information:
Western Blotting (WB), Immunohistochemistry (IHC) and ELISA. Suggested dilutions for WB at 1:500 - 1:1000, ELISA at 1:5,000-10,000 and IHC at 1:50-1:100. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: V5 Tag
Clonality: Monoclonal
Clone: BE10
Conjugation: Unconjugated
Epitope: GKPIPNPLLGLDST
Host: Mouse
Isotype:
Reactivity: