Anti-HA Tag Mouse Monoclonal Antibody [clone: BHAC5]
Catalog # BSENM-1309-100
Supplier: Biosensis
Specifications
- Antibody type:Tag
- Antigen name:HA tag
- Antigen symbol:HA
- Clonality:Monoclonal
- Clone:BHAC5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Epitope:HA-tag YPYDVPDYA
- Cross adsorption:No
- Format:Liquid
- Immunogen:A synthetic peptide from influenza hemagglutinin epitope (YPYDVPDYA) coupled to KLH.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The Human influenza hemagglutin (HA) tag corresponds to a region (98-106 amino acids) from the HA molecule.
Antibody detects HA-tagged proteins (on amino- and carboxy termini) in transfected mammalian cells.
Application Information:
Western Blotting (WB), Immunoprecipitation (IP) and Immunohistochemistry (IH). Suggested dilutions for WB of 1:1000, IP and IS at 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Tag
Antigen: HA Tag
Clonality: Monoclonal
Clone: BHAC5
Conjugation: Unconjugated
Epitope: HA-tag YPYDVPDYA
Host: Mouse
Isotype:
Reactivity: