Order Entry
Austria
ContactUsLinkComponent
Anti-Catenin beta Sheep Polyclonal Antibody
Anti-Catenin beta Sheep Polyclonal Antibody
Catalog # BSENS-1234-100
Supplier:  Biosensis
undefined
Anti-Catenin beta Sheep Polyclonal Antibody
Catalog # BSENS-1234-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Catenin beta
  • Antigen symbol:
    CTNNB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    amino acid region 771-781 of Human Catenin beta
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Catenin beta-1
  • Immunogen:
    A synthetic peptide (C-SNQLAWFDTDL) corresponding to the amino acid region 771-781 of human Catenin beta coupled to carrier protein KLH.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Catenin beta is an adherens junction protein and has a role in the regulation of cell adhesion and in signal transduction through the Wnt pathway. At least 2 isoforms are produced by alternative splicing.

The specificity of this antibody has been confirmed by WB against the antigen. This antibody was also validated for immunoprecipitation in rat liver lysate (data not shown).
Application Information:
Western Blotting (WB) and Immunoprecipitation. A concentration of 1.0 µg/ml is recommended for WB. Human Catenin beta (isoform 1) has a predicted length of 781 residues and MW of 86 kDa. A concentration of 5 µg/500 µg of lysate is recommended for immunoprecipitation. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Catenin beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: amino acid region 771-781 of Human Catenin beta
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat