Order Entry
Austria
ContactUsLinkComponent
Anti-NT3 Rabbit Polyclonal Antibody
Anti-NT3 Rabbit Polyclonal Antibody
Catalog # BSENR-096-500
Supplier:  Biosensis
undefined
Anti-NT3 Rabbit Polyclonal Antibody
Catalog # BSENR-096-500
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurotrophin-3
  • Antigen symbol:
    NT3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Chicken,
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    aa: 139-149), Mouse and rat NT3 protein
  • Cross adsorption:
    No
  • Neutralisation:
    Yes
  • Format:
    Lyophilized
  • Immunogen:
    A synthetic peptide (YAEHKSHRGEY) as part of human (aa: 139-149), mouse and rat NT3 protein conjugated to BSA has been used as the immunogen.
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

FUNCTION: Seems to promotes the survival of visceral and proprioceptive sensory neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family.

A cross reactivity of less than 0.1% to mouse NGF, recombinant human BDNF and NT4/5 has been shown by dot blot.
Application Information:
IHC, ELISA, WB, inhibition of biological activity. A concentration of 1-10 µg/ml is recommended for IHC, ELISA, WB and inhibition of biological activity in vitro; 2-10 µg/ml (ED50) for in vivo use. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa: 139-149), Mouse and rat NT3 protein
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Chicken, Human