Anti-pan synuclein Sheep Polyclonal Antibody
Artikelnr. BSENS-044-100
Lieferant: Biosensis
Spezifikationen
- Antibody type:Primary
- Antigen name:pan synuclein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:aa 11-22 Human alpha-, beta-, and gamma synuclein proteins
- Cross adsorption:No
- Format:Lyophilized
- Immunogen:A synthetic peptide (AKEGVVAAAEKTKQGV) as a consensus part of human alpha-, beta-, and gamma synuclein proteins conjugated to diphteria toxoid has been used as the immunogen.
- Size:100 µg
- Pk:100 µG
Spezifikationen
Über diesen Artikel
NULL
Overlap specific immunohistochemical staining of alpha-, beta- and gamma synucleins
Application Information:
IHC, WB. A concentration of 3 µg/ml is recommended for immunohistochemistry (frozen & paraffin embedded sections) and 1 µg/ml for western blot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: pan synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa 11-22 Human alpha-, beta-, and gamma synuclein proteins
Host: Sheep
Isotype: IgG
Reactivity: Human, Rat