Order Entry
Austria
ContactUsLinkComponent
Anti-Endomorphin-1 and Endomorphin-2 Rabbit Polyclonal Antibody
Anti-Endomorphin-1 and Endomorphin-2 Rabbit Polyclonal Antibody
Catalog # BSENR-1495-50
Supplier:  Biosensis
undefined
Anti-Endomorphin-1 and Endomorphin-2 Rabbit Polyclonal Antibody
Catalog # BSENR-1495-50
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endomorphin-1 and Endomorphin-2
  • Antigen symbol:
    EM 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Immunogen:
    Antibodies to human Endomorphins 1 and 2 were made by immunising rabbits with the human Endomorphin-1 peptide (YPWF) and then selecting those antibodies that cross-reacted with both human Endomorphins 1 and 2.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Endomorphins 1 and 2 are endogenous opioid peptides which have the highest affinity for the mu-opioid receptors. Located in various parts of the brain and interacts with mu-opioid receptors and produces analgesia.

Antibody recognizes Endomorphins. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross-reactivity Human; mouse; rat. Endomorphin-1 is highly conserved molecule so cross-reactivity with other species is expected. Cross-reactivity with other opioid peptides is as follows: with Endomorphin-2 = 70%; Met-enkephalin <0.03%; with Leu-enkephalin 0.01%; with beta-endorphin<0.1%.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Endomorphin-1 and Endomorphin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat