Order Entry
Austria
ContactUsLinkComponent
Anti-Myc Tag Rabbit Polyclonal Antibody
Anti-Myc Tag Rabbit Polyclonal Antibody
Catalog # BSENR-1319-100
Supplier:  Biosensis
undefined
Anti-Myc Tag Rabbit Polyclonal Antibody
Catalog # BSENR-1319-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    MYC Tag
  • Antigen symbol:
    MYC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    Myc epitope
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    Myc epitope tag peptide
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The Myc tag contains the amino acids Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu (E-Q-K-L-I-S-E-E-D-L) corresponding to amino acids 410-419 of human Myc. This tag is widely used for monitoring expression of recombinant proteins in bacteria, insect and mammalian cells.

This polyclonal anti-Myc-tag antibody detects overexpressed proteins containing the Myc epitope tag. The antibody recognizes the Myc-tag fused to either the amino- or carboxy- termini of targeted proteins in transfected or transformed cells.
Application Information:
ELISA and Western Blotting (WB). Suggested dilutions for ELISA at 1:1,000 and WB 1:500 - 1:1,000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: Myc Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Myc epitope
Host: Rabbit
Isotype:
Reactivity: