Oxidase test strips
Frequently Bought Together
Test tube racks
: Brand
Disinfectant liquid, terralin® protect
: SCHÜLKE & MAYR GMBH
Forceps
: RSG
Tough-Tags® label rolls
: DIVERSIFIED BIOTECH
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
BioSart® 100 monitors
: Sartorius
L(+)-Arabinose 99+%
: Thermo Fisher Scientific
Peqlab, Strips of 8 PCR Tubes
: VWR Peqlab
Potassium hydroxide 3% in ethanol 95% Reag. Ph. Eur. 1070303
: VWR Chemicals
Methylene blue 1% (w/v) in aqueous solution
: Thermo Fisher Scientific
Demeter-Pipette 1,1ml 1 * 1.000 items
: SARSTEDT
VWR® Culture Media, Ready To Use, In Plates
: VWR Chemicals
VWR®, LB media, pre-mixed
: VWR Chemicals
Lids for Aqua Pro range shaking water baths
: GRANT INSTRUMENTS