ContactUsLinkComponent
384-well microplates, ShallowWell, standard height
384-well microplates, ShallowWell, standard height
Catalog # 734-2876
   Low Manufacturing Impact
384-well microplates, ShallowWell, standard height
Catalog # 734-2876
   Low Manufacturing Impact
Specifications
  • Recommended working volume:
    2 - 20 µl
  • Colour:
    Clear
  • Description:
    384 ShallowWell plate, non treated surface, without lid
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    100
  • Well volume:
    25 µl
Specifications
About this item
These standard height plates are suitable for automation, without the need to make adjustments to robotics, plate readers or liquid handlers.
  • Untreated hydrophobic polystyrene suited for growth of suspension cultures
  • ANSI/SBS standard footprint