Millex® Syringe Filters, PTFE Membrane
: Merck
Syringe filters with hydrophobic PTFE membrane. Used in vacuum line protection, transducer protection and gas filtration. Available as either 25 mm or pump-driven Millex®-FG₅₀ 50 mm diameter filters.
Filters are either non sterile or sterilised with ethylene oxide.
- Catalog No:
- 516-6551
- MILFSLFG75000
- MILFSLFG75010
- MILFSLFA05000
- MILFSLFG0250M
- MILFSLFG05010
- MILFSLFG65000
- MILFSLFH05000
- 514-0410
- MILFSLFG05000
- MILFSLFH05010
- MILFSLFA05010
- 514-0409
- MILFSLFGL25BS
- MILFSLFG65010
- MILFSLFGN25VS
- Max. operating pressure:
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 5,2 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 2,0 bar
- Max. temperature:
- 121 °C
- 121 °C
- 121 °C
- 121 °C
- 45 °C
- 130 °C
- 130 °C
- 121 °C
- 121 °C
- 130 °C
- 121 °C
- 121 °C
- 121 °C
- 121 °C
- 130 °C
- 45 °C
Frequently Bought Together