Orders LinkContactUsLinkComponent
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution
Supplier:  VWR Chemicals
Certificates
VWR®NucleasEliminator™, Decontamination solution
VWR®NucleasEliminator™, Decontamination solution
3 Options Available Below
Product Details & Documents
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.
  • Effectively removes and deactivates nuclease contamination, even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years
Simply wipe, immerse, or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature, and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips, and electrophoresis units.
A 100 ml bottle of the ready to use liquid will treat up to 100 microcentrifuge tubes or 25 casting trays for agarose electrophoresis.
Product Family Options
NucleasEliminator™ ready to use liquid
Product Information
  • Pk
    Availability
    Price
  • E891-100ML
    NucleasEliminator™, ready to use liquid
    NucleasEliminator™, ready to use liquid
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 39,00 €
    39,00 €
    1 * 100 mL
       39,00 €
  • E891-500ML
    NucleasEliminator™, ready to use liquid
    NucleasEliminator™, ready to use liquid
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 141,00 €
    141,00 €
    1 * 500 mL
       141,00 €
NucleasEliminator™ wipes
Product Information
  • Pk
    Availability
    Price
  • E891-25PK
    NucleasEliminator™, impregnated wipes
    NucleasEliminator™, impregnated wipes
    25
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 90,30 €
    90,30 €
    1 * 25 ST
       90,30 €