Wound dressing kit, QuickStop
118-0549
- Description:Wound dressing kit, QuickStop, with one large wound dressing (17×17 cm) and two small wound dressings (12×7,5 cm)
- Environmentally Preferable:
- Pk:3
Wound dressing kit for effective first aid of both minor and heavy bleeding. For heavy bleeding, such as the rupture of a main artery, the small wound dressing can be put into the pocket of the large wound dressing to function as an additional compression.
- Can be used separately or for restocking QuickSafe stations
- Kit replaces a number of traditional dressings
: The kit contains one wound dressing with compression (17×17 cm) and two small wound dressings with compression (12×7,5 cm)