Anti-AMY1C Mouse Polyclonal Antibody
ABNOH00000278-B01P
: Abnova
- Antibody type:Primary
- Antigen name:AMY1C
- Antigen symbol:AMY1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:278
- Antigen synonyms:AMY1
- Amino acid number:1 to 511
- Storage buffer:1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:AMY1C (AAI56581.1, 1 a.a. ~ 511 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Mouse polyclonal antibody raised against a full-length human AMY1C protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: AMY1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human