Anti-ATG16L1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13324
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Autophagy Related 16-like 1
- Antigen symbol:ATG16L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:55054
- Antigen synonyms:IBD10|APG16-like 1|WDR30|Autophagy-related protein 16-1|APG16L|ATG16L1
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region 18 amino acids of human ATG16L1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ATG16L1.
Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml
Type: Primary
Antigen: ATG16L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat