Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 4H11[APG]]
ABNOMAB4412
: Abnova
- Antibody type:Primary
- Antigen name:CD34 molecule
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:4H11[APG]
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:947
- Antigen synonyms:CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD34 from permanent human cell line derived from peripheral leucocytes of a patient suffering from chronic myeloid leukaemia.
- Size:100 Reactions
- Pk:100 µg
Mouse monoclonal antibody raised against native CD34.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11[APG]
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human