Anti-AES Rabbit Polyclonal Antibody
ABNOPAB13165
: Abnova
- Antibody type:Primary
- Antigen name:Amino-terminal Enhancer of Split
- Antigen symbol:AES
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:166
- Antigen synonyms:AES-1|AES-2|TLE5|Grg-5|ESP1|GRG5|GRG
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 16 amino acids of human AES.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of AES.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: AES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat