Anti-CRABP1 Mouse Monoclonal Antibody [clone: AT1A1]
ABNOMAB7558
: Abnova
- Antibody type:Primary
- Antigen name:Cellular retinoic acid binding protein 1
- Antigen symbol:CRABP1
- Clonality:Monoclonal
- Clone:AT1A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1381
- Antigen synonyms:CRABPI|CRABP|RBP5|CRABP-I
- Amino acid number:1 to 137
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-137 of human CRABP1.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against recombinant CRABP1.
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: CRABP1
Clonality: Monoclonal
Clone: AT1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human