Anti-PTPRC Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: EM-05]
ABNOMAB5118
: Abnova
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:EM-05
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG
- Reactivity:Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:19264
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified Ptprc from mouse peripheral blood leukocytes.
- Size:100 μg
- Pk:100 µg
Rat monoclonal antibody raised against native Ptprc.
Recommended Dilutions: Flow Cytometry: 4 ?g/ml
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: EM-05
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse