Anti-CSDE1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB15731
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cold Shock Domain Containing E1
- Antigen symbol:CSDE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:229663
- Antigen synonyms:BC016898|AA960392|mKIAA0885|unr|D3Jfr1
- Storage buffer:PBS (50% glycerol, 0,02% sodium azide)
- Sequence:QNAQTMAYNITPLRRATVECVKDQFGFINYEVGDSKKLFFHVKEVQDGVELQAGDEVEFSVILNQRTGKCSACNVWRVCEGPKAVAAPRPDRLVNRLKNITLDDASAPRLMVLRQPRGPDNSMGFGAERKIRQAGVID
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 138 mouse Csde1.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant Csde1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CSDE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human