Order Entry
Austria
ContactUsLinkComponent
Mouse IgG1 Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex))
Mouse IgG1 Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # 67-4714-U100
Supplier:  Tonbo Biosciences
Mouse IgG1 Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex))
Catalog # 67-4714-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Cross adsorption:
    No
  • Storage temperature:
    Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Pk:
    100 µG

Specifications

About this item

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: