Order Entry
Austria
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: L17F12]
Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: L17F12]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: L17F12]
Supplier:  Tonbo Biosciences
Certificates

Specifications

  • Catalog No:
  • 35-0058-T500
  • 35-0058-T025
  • 35-0058-T100
  • Antigen symbol:
  • CD5
  • CD5
  • CD5
  • Antigen name:
  • CD5 molecule
  • CD5 molecule
  • CD5 molecule
  • Antigen synonyms:
  • CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
  • CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
  • CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • L17F12
  • L17F12
  • L17F12
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 921
  • 921
  • 921
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together