Rat IgG1 Isotype Control [clone: HRPN] (violetFluor® 450)
Catalog # 75-4301-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clone:HRPN
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:
- Environmentally Preferable:
- Neutralisation:Yes
- Storage temperature:Store Undiluted at 2 to 8 °C
- Size:100 μg
- Pk:100 µG
Specifications
About this item
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.
Type: Control
Antigen: HRPN
Clonality:
Clone: HRPN
Conjugation: violetFluor™ 450
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: N/A