Rat IgG2a Isotype Control [clone: 2A3]
40-4321-U100
- Antibody type:Control
- Clone:2A3
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Neutralisation:Yes
- Storage temperature:Store undiluted at 2-8 °C, protect from light, do not freeze.
- Size:100 μg
- Pk:100 µG
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.
Type: Control
Antigen: 2A3
Clonality:
Clone: 2A3
Conjugation: In Vivo Ready™
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: