Order Entry
Austria
ContactUsLinkComponent
Anti-CD19 Rat Antibody (redFluor™ 710) [clone: 1D3]
Anti-CD19 Rat Antibody (redFluor™ 710) [clone: 1D3]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD19 Rat Antibody (redFluor™ 710) [clone: 1D3]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 80-0193-U100
  • 80-0193-U025
  • Antigen symbol:
  • CD19
  • CD19
  • Antigen name:
  • CD19 molecule
  • CD19 molecule
  • Antigen synonyms:
  • B-lymphocyte antigen CD19. B4|CVID3|AW495831|mouse CD19|Cluster of Differentiation 19
  • B-lymphocyte antigen CD19. B4|CVID3|AW495831|mouse CD19|Cluster of Differentiation 19
  • Antibody type:
  • Primary
  • Primary
  • Conjugation:
  • redFluor® 710
  • redFluor® 710
  • Clone:
  • 1D3
  • 1D3
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12478
  • 12478
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store Undiluted at 2 to 8 °C
  • Store Undiluted at 2 to 8 °C

Specifications

Related Information

Frequently Bought Together