Order Entry
Austria
ContactUsLinkComponent
Anti-CD11c Mouse Antibody (redFluor™ 710) [clone: 3.9]
Anti-CD11c Mouse Antibody (redFluor™ 710) [clone: 3.9]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD11c Mouse Antibody (redFluor™ 710) [clone: 3.9]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 80-0116-T025
  • 80-0116-T100
  • Antigen symbol:
  • ITGA10
  • ITGA10
  • Antigen name:
  • Integrin Alpha 10
  • Integrin Alpha 10
  • Antigen synonyms:
  • integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
  • integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
  • Antibody type:
  • Primary
  • Primary
  • Conjugation:
  • redFluor® 710
  • redFluor® 710
  • Clone:
  • 3.9
  • 3.9
  • Reactivity:
  • Marmoset, Capuchin monkey, Chimpanzee, Human
  • Marmoset, Capuchin monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3687
  • 3687
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together