These sterile syringe filters are ideal for sterilising (0,2 µm) or clarifying biological solutions, such as cell culture and microbial media, reagents and additives, proteinaceous solutions and buffers.
- Three-year shelf life, with lot number and expiration date printed on individual syringe filter packages for easy traceability
- Colour coded, with membrane and pore size printed on the syringe filter
- Female Luer lock/male Luer slip inlet/outlet connection
PES membrane; PP housing: For sterile filtration and clarification of cell culture media, buffers and additives. Higher flow rates, lower protein binding and lower extractables than cellulosic membranes. Naturally hydrophilic; contains no wetting agents. Certified sterile, non-cytotoxic and non-pyrogenic.
SFCA membrane; PP housing: For cell culture fluids and cellular solutions. Low protein binding, lower extractables than standard CA.
CA membrane; PP housing: For sterile filtration and clarification of buffers, aqueous solutions and reagents. Some protein binding and extractables.
Nylon membrane; PP housing: Useful for alcohols and weak solvents. Naturally hydrophilic, low extractables, some protein binding.
CA membrane and integral glass fibre pre-filter, PP housing: These syringe filters are designed for filtering high particulate or viscous suspensions or solutions. Filter and pre-filter in one disposable unit. Certified non-cytotoxic and non-pyrogenic.
- Catalog No:
- 516-0930
- 516-0945
- 516-0946
- 516-0944
- 516-0935
- 516-0939
- 516-0936
- 516-0934
- 516-0943
- 516-0937
- 516-0938
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement