Anti-GPR41 Rabbit Polyclonal Antibody
Catalog # CAYM16363-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:GPR41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:No
- Storage buffer:TBS, pH 7,4, conaining 50% glycerol, 0,1% BSA, and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: GPR41
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: Internal
Host: Rabbit
Isotype: IgG
Reactivity: Human