Order Entry
Austria
ContactUsLinkComponent
Anti-MPO Mouse Monoclonal Antibody [clone: 2C8]
Anti-MPO Mouse Monoclonal Antibody [clone: 2C8]
Catalog # CAYM15638-1
Supplier:  Cayman Chemical
Anti-MPO Mouse Monoclonal Antibody [clone: 2C8]
Catalog # CAYM15638-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MPO
  • Clonality:
    Monoclonal
  • Clone:
    2C8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Mouse
  • Cross adsorption:
    No
  • Storage buffer:
    PBS, pH 7,2, containing 50% glycerol, 0,5 mg/ml BSA, and 0,02% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: 2C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse