Anti-dsDNA Mouse Monoclonal Antibody [clone: 2C4]
CAYM15635-1
- Antibody type:Primary
- Antigen symbol:dsDNA
- Clonality:Monoclonal
- Clone:2C4
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human,Mouse
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2, containing 50% glycerol, 0,5 mg/ml BSA, and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:ammonium sulfate purified
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: dsDNA
Clonality: Monoclonal
Clone: 2C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse