Anti-Abeta42 Mouse Monoclonal Antibody [clone: 6C3, MOAB-2]
CAYM11610-200
- Antibody type:Primary
- Antigen name:Amyloid beta 1-42
- Clonality:Monoclonal
- Clone:6C3, MOAB-2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2, containing 50% glycerol, and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:oligomeric form of A-beta42
- Purification:purified
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: Abeta42
Clonality: Monoclonal
Clone: 6C3, MOAB-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human