Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RIV7]
CAYM10010530-1
- Antibody type:Primary
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:RIV7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Storage buffer:PBS, pH 7,2
- Storage temperature:+4 °C
- Shipping temperature:4 °C
- Purification:purified
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RIV7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human