Anti-PTEN Rabbit Polyclonal Antibody
CAYM10005059-500
- Antibody type:Primary
- Antigen symbol:PTEN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:254 to 270
- Sequence:KVEFFHKQNKMLKKDKM
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human PTEN amino acids 254-270
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Chimpanzee, Human, Mouse, Rat