Anti-GABRA6 Rabbit Polyclonal Antibody
ROCK600-401-D48
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, alpha 6
- Antigen symbol:GABRA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:29708
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-GABA(A) Receptor alpha 6 Antibody was produced in rabbit by repeated immunizations with recombinant fusion Proteins from the cytoplasmic loop of the alpha 6 subunit.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Anti-GABA(A) Receptor alpha 6 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 57 kDa in size corresponding to the alpha 6 subunit of the GABA(A) Receptor.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GABRA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat