Anti-GABRA3 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-D46
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, alpha 3
- Antigen symbol:GABRA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:24947
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-GABA(A) Receptor alpha 3 Antibody was produced by repeated immunizations with a fustion Protein from the N-terminal region of the alpha 3 subunit.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-GABA(A) Receptor alpha 3 (Rabbit) antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 51 kDa in size corresponding to the alpha 3 subunit of the GABA(A) Receptor.
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: GABRA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat